2-噻吩乙胺 ≥98.0% (by GC)
供应商: TCI
|
Danger
|
Synonyms:
2-(Thiophen-2-yl)ethanamine, 2-(2-Aminoethyl)thiophene
A1637-25G
A1637-250G
TCIAA1637-25GEA
0
CNY
TCIAA1637-25G
TCIAA1637-250G
2-噻吩乙胺 ≥98.0% (by GC)
2-噻吩乙胺
分子式:
C₆H₉NS 分子量: 127.21 g/mol 沸点: 201°C 闪点: 88°C |
MDL编号:
MFCD00051495 CAS号: 30433-91-1 EINECS: 250-196-9 UN号: 2735 ADR: 8 II |
|
产品技术规格
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...