2,6-二氯吡啶 N-氧化物 98%

供应商: Thermo Fisher Scientific
Warning

Synonyms: 2,6-Dichloropyridine 1-oxide

379420010 379420050
ACRO379420010EA 0 CNY
ACRO379420010 ACRO379420050
2,6-二氯吡啶 N-氧化物 98%
2,6-二氯吡啶 N-氧化物
分子式: C₅H₃Cl₂NO
分子量: 163.99 g/mol
溶点: 134…138 °C
存储条件: Ambient
MDL编号: MFCD00130246
CAS号: 2587-00-0
UN号: 2811
ADR: 6.1 III

现在订购

产品技术规格

Appearance White to cream to light yellow Powder or crystals
Infrared spectrum Conforms
Melting point 134 to 142 °C
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR