2-Chloro-6-hydroxynicotinic acid ≥97.0% (by GC, titration analysis)

供应商: TCI
Warning

C2398-1G C2398-5G
TCIAC2398-1GEA 0 CNY
TCIAC2398-1G TCIAC2398-5G
2-Chloro-6-hydroxynicotinic acid ≥97.0% (by GC, titration analysis)
2-Chloro-6-hydroxynicotinic acid
分子式: C₆H₄ClNO₃
分子量: 173.5500 g/mol
溶点: 302°C
MDL编号: MFCD09757508
CAS号: 38025-90-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=912c0f0f73fd21f48e8f2ff46c3b373b8b752318&vid=912c0f0f73fd21f48e8f2ff46c3b373b8b752318&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Light yellow to Light red powder to crystal
Purity (GC) min. 97.0 %
Purity (Neutralization titration) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR