1,4-Bis(bromomethyl)naphthalene >65.0% (通过GC) (contains isomer)

供应商: TCI
Danger

B1919-1G B1919-5G
TCIAB1919-1GEA 0 CNY
TCIAB1919-1G TCIAB1919-5G
1,4-Bis(bromomethyl)naphthalene >65.0% (通过GC) (contains isomer)
1,4-Bis(bromomethyl)naphthalene
分子式: C₁₂H₁₀Br₂
溶点: 169…173 °C
MDL编号: MFCD01321143
CAS号: 58791-49-4
UN号: 3261
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=381f37d8d9e805c16172aa5198ddc95c1c5da28e&vid=381f37d8d9e805c16172aa5198ddc95c1c5da28e&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (GC) min. 65.0 %
Purity (Argentmetric Titration) min. 98.0 %
Melting point 169.0 to 173.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR