1-Benzyl-1-phenylhydrazine hydrochloride ≥98.0% (by titrimetric analysis)

供应商: TCI
Warning

Synonyms: N-Benzyl-N-phenylhydrazine hydrochloride

B0434-25G B0434-500G
TCIAB0434-25GEA 0 CNY
TCIAB0434-25G TCIAB0434-500G
1-Benzyl-1-phenylhydrazine hydrochloride ≥98.0% (by titrimetric analysis)
1-Benzyl-1-phenylhydrazine hydrochloride
分子式: C₁₃H₁₅ClN₂
分子量: 234.73 g/mol
MDL编号: MFCD00050690
CAS号: 5705-15-7
EINECS: 227-195-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e69f68469dc8d3a36c94d447342cd1868dea51fb&vid=e69f68469dc8d3a36c94d447342cd1868dea51fb&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) min. 98.0 %
Melting point 174.0 to 178.0 deg-C
Solubility in EtOH almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR