5-乙酰水杨酸甲酯 ≥98.0% (通过GC)

供应商: TCI
Warning

M1658-25G M1658-500G
TCIAM1658-25GEA 0 CNY
TCIAM1658-25G TCIAM1658-500G
5-乙酰水杨酸甲酯 ≥98.0% (通过GC)
5-乙酰水杨酸甲酯
分子式: C₁₀H₁₀O₄
沸点: 167°C
MDL编号: MFCD00040924
CAS号: 16475-90-4
EINECS: 240-532-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dd581240829d8f85dfbede0e51797e13193e2df8&vid=dd581240829d8f85dfbede0e51797e13193e2df8&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Light yellow to Dark green powder to crystal
Purity (GC) min. 98.0 %
Melting point 58.0 to 62.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR