4,4'-Diiodo-trans-stilbene ≥97.0% (通过GC)

供应商: TCI
Warning

D3094-1G D3094-5G
TCIAD3094-1GEA 0 CNY
TCIAD3094-1G TCIAD3094-5G
4,4'-Diiodo-trans-stilbene ≥97.0% (通过GC)
4,4'-Diiodo-trans-stilbene
分子式: C₁₄H₁₀I₂
CAS号: 201861-91-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=208600348aa53c266c99f665e9308e8f8cfc862d&vid=208600348aa53c266c99f665e9308e8f8cfc862d&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR