乙醇酸乙酯 95%

供应商: Thermo Fisher Scientific
Warning

Synonyms: Ethyl Hydroxyacetate, Hydroxyacetic Acid Ethyl Ester, Glycolic Acid Ethyl Ester, Ethyl 2-hydroxyacetate

410080250 410081000
ACRO410080250EA 0 CNY
ACRO410080250 ACRO410081000
乙醇酸乙酯 95%
乙醇酸乙酯
分子式: C₄H₈O₃
沸点: 158…159 °C (760 mmHg)
密度: 1,08 g/cm³
闪点: 62 °C
存储条件: Ambient
MDL编号: MFCD00021970
CAS号: 623-50-7
EINECS: 210-798-4

现在订购

产品技术规格

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4170 to 1.4200 (20°C, 589 nm)
GC ≥94 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR