三甲基硅乙酸酯 ≥98.0% (by GC)

供应商: TCI
Danger

Synonyms: Trimethylsilyl acetate

A1567-25ML A1567-250ML
TCIAA1567-25MLEA 0 CNY
TCIAA1567-25ML TCIAA1567-250ML
三甲基硅乙酸酯 ≥98.0% (by GC)
三甲基硅乙酸酯
分子式: C₅H₁₂O₂Si
闪点: 4°C
MDL编号: MFCD00008695
CAS号: 2754-27-0
EINECS: 220-404-2
UN号: 1993
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9a9de1b89c9f5ac28973daf6ee665d658b5daa25&vid=9a9de1b89c9f5ac28973daf6ee665d658b5daa25&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR