6-氟-4-氧代-4氢-1-苯并吡喃-2-羧酸 ≥97.0% (by HPLC, titration analysis)

供应商: TCI
Warning

Synonyms: 6-Fluoro-4-oxo-4H-chromene-2-carboxylic acid

F0744-25G F0744-5G
TCIAF0744-25GEA 0 CNY
TCIAF0744-25G TCIAF0744-5G
6-氟-4-氧代-4氢-1-苯并吡喃-2-羧酸 ≥97.0% (by HPLC, titration analysis)
6-氟-4-氧代-4氢-1-苯并吡喃-2-羧酸
分子式: C₁₀H₅FO₄
分子量: 208.15 g/mol
溶点: 258°C
MDL编号: MFCD03070543
CAS号: 99199-59-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4f7792246ef1572cd252c4715eebede8b8d2d8cf&vid=4f7792246ef1572cd252c4715eebede8b8d2d8cf&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (Neutralization titration) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR