1-Benzyl-4-cyano-4-hydroxypiperidine hydrochloride ≥98.0% (by HPLC, titration analysis)

供应商: TCI
Danger

Synonyms: 1-Benzyl-4-hydroxypiperidine-4-carbonitrile hydrochloride

B2237-25G B2237-5G
TCIAB2237-25GEA 0 CNY
TCIAB2237-25G TCIAB2237-5G
1-Benzyl-4-cyano-4-hydroxypiperidine hydrochloride ≥98.0% (by HPLC, titration analysis)
1-Benzyl-4-cyano-4-hydroxypiperidine hydrochloride
分子式: C₁₃H₁₇ClN₂O
MDL编号: MFCD00012836
CAS号: 71617-20-4
EINECS: 275-713-5
UN号: 3439
ADR: 6.1 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1a24fa4de6b6bed29871102967f25969b80d9836&vid=1a24fa4de6b6bed29871102967f25969b80d9836&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Argentmetric Titration) min. 98.0 %
Solubility in Water very faint turbidity


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR