环己烷丙酸烯丙酯 ≥98.0% (by GC)
供应商: TCI
|
Warning
|
Synonyms:
Allyl Cyclohexanepropionate, Cyclohexanepropanoic acid 2-propen-1-yl ester, Allyl 3-cyclohexylpropanoate, 2-Propenyl 3-cyclohexylpropanoate, Cyclohexanepropionic acid allyl ester, Allyl 3-cyclohexylpropionate, 3-Cyclohexylpropionic acid Allyl Ester Cyclohexanepropionic acid Allyl Ester
C0485-25ML
C0485-500ML
TCIAC0485-25MLEA
0
CNY
TCIAC0485-25ML
TCIAC0485-500ML
环己烷丙酸烯丙酯 ≥98.0% (by GC)
环己烷丙酸烯丙酯
Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: Cyclohexanepropionic Acid Allyl Ester, 3-Cyclohexylpropionic Acid Allyl Ester
分子式:
C₁₂H₂₀O₂ 分子量: 196.29 g/mol 沸点: 91°C 闪点: 126°C |
MDL编号:
MFCD00059557 CAS号: 2705-87-5 EINECS: 220-292-5 |
|
产品技术规格
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Solubility in Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...