9,10-Anthraquinone-2-carbonyl chloride ≥98.0% (by HPLC, titration analysis)

供应商: TCI
Danger

Synonyms: 9,10-Dioxo-9,10-dihydroanthracene-2-carbonyl chloride, Anthraquinone-2-carbonyl Chloride

A0503-25G A0503-5G
TCIAA0503-25GEA 0 CNY
TCIAA0503-25G TCIAA0503-5G
9,10-Anthraquinone-2-carbonyl chloride ≥98.0% (by HPLC, titration analysis)
9,10-Anthraquinone-2-carbonyl chloride
分子式: C₁₅H₇ClO₃
分子量: 270.67 g/mol
沸点: 210°C
MDL编号: MFCD00059498
CAS号: 6470-87-7
UN号: 3261
ADR: 8 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1ae3602681b6e5ff21ba792bfb8d30f5d8acac8c&vid=1ae3602681b6e5ff21ba792bfb8d30f5d8acac8c&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Light orange to Yellow to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Argentmetric Titration) min. 98.0 %
Melting point 146.0 to 150.0 deg-C
Solubility in hot Toluene almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR