4,4'-Dinitrostilbene ≥98.0% (通过HPLC)

供应商: TCI

D2464-25G D2464-5G
TCIAD2464-25GEA 0 CNY
TCIAD2464-25G TCIAD2464-5G
4,4'-Dinitrostilbene ≥98.0% (通过HPLC)
4,4'-Dinitrostilbene
分子式: C₁₄H₁₀N₂O₄
溶点: 295°C
MDL编号: MFCD00017043
CAS号: 2501-02-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=707491b8f0d4448df31bc1592bb502042ec4bfe5&vid=707491b8f0d4448df31bc1592bb502042ec4bfe5&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Yellow to Amber powder to crystal
Purity (HPLC) min. 98.0 area%
Solubility in hot DMF almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR