3-Bromo-2,4-dimethyl-5-phenylthiophene ≥98.0% (by GC)

供应商: TCI

B2677-1G B2677-5G
TCIAB2677-1GEA 0 CNY
TCIAB2677-1G TCIAB2677-5G
3-Bromo-2,4-dimethyl-5-phenylthiophene ≥98.0% (by GC)
3-Bromo-2,4-dimethyl-5-phenylthiophene
分子式: C₁₂H₁₁BrS
存储条件: Refrigerator
MDL编号: MFCD08276302
CAS号: 362513-28-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9f06cacc6d83e3238b8f42a66f9bdec55541dca4&vid=9f06cacc6d83e3238b8f42a66f9bdec55541dca4&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Orange to Green powder to lump
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR