4-溴-1,8-萘二甲酸酐 ≥95.0% (by titrimetric analysis)

供应商: TCI
Warning

Synonyms: 6-Bromobenzo[de]isochromene-1,3-dione

B0858-25G B0858-5G
TCIAB0858-25GEA 0 CNY
TCIAB0858-25G TCIAB0858-5G
4-溴-1,8-萘二甲酸酐 ≥95.0% (by titrimetric analysis)
4-溴-1,8-萘二甲酸酐
分子式: C₁₂H₅BrO₃
分子量: 277.0703 g/mol
MDL编号: MFCD00006927
CAS号: 81-86-7
EINECS: 201-382-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f99436a82745d1925b86328bc604b93c6295568c&vid=f99436a82745d1925b86328bc604b93c6295568c&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Light yellow powder to crystaline
Purity (Methanolysis Method) min. 95.0 %
Melting point 218.0 to 224.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR