1-(2-Furoyl)piperazine hydrochloride ≥98.0% (by HPLC, total nitrogen)

供应商: TCI
Warning

Synonyms: Furan-2-yl(piperazin-1-yl)methanone hydrochloride

F0705-25G F0705-5G
TCIAF0705-25GEA 0 CNY
TCIAF0705-25G TCIAF0705-5G
1-(2-Furoyl)piperazine hydrochloride ≥98.0% (by HPLC, total nitrogen)
1-(2-Furoyl)piperazine hydrochloride
分子式: C₉H₁₃ClN₂O₂
分子量: 216.6647 g/mol
MDL编号: MFCD00236012
CAS号: 60548-09-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fa0c04857ea086654ec1be03ba961dc3f7004f62&vid=fa0c04857ea086654ec1be03ba961dc3f7004f62&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 204.0 to 209.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR