1-(2-Tetrahydrofuroyl)piperazine 98%

供应商: Thermo Fisher Scientific
Danger

Synonyms: (Piperazin-1-yl)(tetrahydrofuran-2-yl)methanone

367160050 367160010
ACRO367160050EA 0 CNY
ACRO367160050 ACRO367160010
1-(2-Tetrahydrofuroyl)piperazine 98%
1-(2-Tetrahydrofuroyl)piperazine
分子式: C₉H₁₆N₂O₂
分子量: 184.24 g/mol
沸点: 140 °C (2mbar)
密度: 1.17 g/cm³
存储条件: Ambient
MDL编号: MFCD00190247
CAS号: 63074-07-7
UN号: 3267
ADR: 8 III

现在订购

产品技术规格

Appearance Colorless to yellow Viscous liquid
Infrared spectrum Conforms
Refractive index 1.5195 to 1.5215 (20°C, 589 nm)
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR