3-Ethyl-2-methylbenzothiazolium iodide ≥98.0% (通过HPLC,滴定分析)
供应商: TCI
|
Warning
|
E0476-25G
E0476-5G
TCIAE0476-25GEA
0
CNY
TCIAE0476-25G
TCIAE0476-5G
3-Ethyl-2-methylbenzothiazolium iodide ≥98.0% (通过HPLC,滴定分析)
3-Ethyl-2-methylbenzothiazolium iodide
分子式:
C₁₀H₁₂INS |
MDL编号:
MFCD00044134 CAS号: 3119-93-5 EINECS: 221-494-6 |
|
产品技术规格
Appearance | White to Dark green powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...