双(六氟乙酰丙酮)合铜(II) ≥95.0% (by titrimetric analysis)

供应商: TCI
Warning

Synonyms: Hexafluoroacetylacetono Copper(II) Salt, Copper(II) Hexafluoroacetylacetonate, Copper(II) hexafluoro-2,4-pentanedionate hydrate, Bis(hexafluoroacetylacetonato)copper(II) hydrate, Copper(II) hexafluoro-2,4-pentanedionate xhydrate

H0554-1G H0554-5G
TCIAH0554-1GEA 0 CNY
TCIAH0554-1G TCIAH0554-5G
双(六氟乙酰丙酮)合铜(II) ≥95.0% (by titrimetric analysis)
双(六氟乙酰丙酮)合铜(II)
分子式: C₁₀H₂CuF₁₂O₄·xH₂O
分子量: 477.64 g/mol (anhy)
MDL编号: MFCD00151019
CAS号: 14781-45-4
EINECS: 238-850-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6300729a5401cc64b6484c4d937aded5960636d8&vid=6300729a5401cc64b6484c4d937aded5960636d8&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Green to Dark green powder to crystal
Purity (Chelometric Titration) min. 95.0 % (calcd.on anh.substance)
Water max. 10.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR