Plasma freezers, Revco®
供应商: REVCO TECHNOLOGIES
UFP1230V UFP430V
REVCUFP1230VEA 0 CNY
REVCUFP1230V REVCUFP430V
Plasma freezers, Revco®
冷柜 实验室通用冰箱
These freezers are designed to meet AABB, ANRC and FDA standards for safety and performance in plasma storage. Five models are available with different cabinet sizes to meet a variety of space needs and storage volume requirements.
  • Simplified operation and increased accuracy with the microprocessor control
  • CFC free refrigerant
  • Preset temperature at −30 °C
  • Positive airflow systems
Automatic condensate removal and defrost, with sensor that automatically initiates defrost cycle on demand, eliminating need for a defrost timer, minimizing frost build-upMore Product Information