Blood Bank Refrigerators
供应商: REVCO TECHNOLOGIES
REB2304V REB404V REB5004V REB1204V REB3004V
REVCREB2304VEA 0 CNY
REVCREB2304V REVCREB404V REVCREB5004V REVCREB1204V REVCREB3004V
Blood Bank Refrigerators
冰箱
These blood bank refrigerators store whole blood and blood components.
  • Second cabinet temperature probe
  • Keyed on/off switch with setpoint security
  • Built-in chart recorder and remote alarm contacts
  • Heat-free defrost for maximum temperature uniformity
  • Scratch-resistant painted chamber and cabinet material
  • Bright LED interior lighting and optional 4 to 20 mA output
The features include microprocessor control systems, positive airflow systems, industrial-quality cabinet construction with adjustable levelling feet and extra-strength refrigerationMore Product Information
现在订购
规格
型号 REB-2304V REB-404V REB-5004V REB-1204V REB-3004V
Environmentally Preferable
货号 REVCREB2304V REVCREB404V REVCREB5004V REVCREB1204V REVCREB3004V
Air circulation 通风 通风 通风 通风 通风
显示 Digital Digital Digital Digital Digital
Defrost Automatic Automatic Automatic Automatic Automatic
报警器 Audible and visual Audible and visual Audible and visual Audible and visual Audible and visual
进入孔 1 1 1 1 1
Shelves/drawers 6 shelves 2 shelves 12 shelves 5 shelves 6 shelves
门型 1 glass swing door 1 solid swing door 2 glass swing doors 1 glass swing door 1 glass swing door
门锁 Yes Yes Yes Yes Yes
Material
重量 213 kg 96 kg 359 kg 145 kg 227 kg