Masterflex® Port Disc Threaded Port with 38-400 Thread Finish Engage 8402+C3156 Cleanroom Manufactured; 10 pack
供应商: MASTERFLEX PROPRIETARY
VWR Item#MFLA40331-05
Your Organization's Price:
正在获取
Large opening makes it suitable for high viscosity liquid and powder applications.
  • 38-400 1" Bottle Cap
  • 2-1/4" disc diameter
Masterflex® Single-Use Bag Ports are engineered to be thermal welded or ultrasonic welded to medical and bioreactor bags of similar material. These materials include polyethylene. These bags can be used to transfer fluid, drug compounds, and cell culture in the biopharmaceudical markets. If material validation is needed, please reach out to our techincal support representatives.