Masterflex® Fittings, Stainless Steel, Elbow, Hose Barb Union, 1/2" ID
供应商: MASTERFLEX PROPRIETARY
VWR Item#MFLA30620-18
Your Organization's Price:
正在获取
Quick and easy assembly without tools.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimise downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
  • Connection Type: 1/2" ID
Create 90° bends in flexible tubing—without the kinks!