Masterflex® Variable-Area Flowmeter with High-Res Valve, Correlated Reading, PTFE Housing and Fittings, 65-mm Scale; 2704 mL/min Air
供应商: MASTERFLEX PREMIUM
VWR Item#MFLB32044-80
Your Organization's Price:
正在获取