ITT Jabsco Polypropylene Pulse Dampeners With Santoprene Diaphragm, 3/8" NPT
供应商: MASTERFLEX PREMIUM
Create a pulse-free flow out of your positive displacement pump.
- Choose between PTFE or Polypropylene with Santoprene dampners
Dampeners work by pre-filling the chamber with compressed air or other gas, then as fluid pressure increases it compresses the air/gas in the chamber. When the air/gas pressure exceeds the fluid pressure the fluid is forced back out of the dampener in between pulse cycles, resulting in a near pulseless flow.<br /><br /><B>More Info:</B> These pulse dampeners can be used with any positive displacement pump that needs flow with reduced pulsation.