John Guest Threaded Elbow Adapters, 3/8" OD to 1/4" NPT(M), PP, 10/Pk
供应商: MASTERFLEX PREMIUM
VWR Item#MFLB34007-64
Your Organization's Price:
正在获取
Quick, leakproof connection.
  • Polypropylene and EPDM materials meet NSF 51, and NSF 61 requirements for food-grade applications; and is FDA compliant
Push-to-connect fittings offer secure connections and quick disconnection. Simply push tubing past the O-ring to the internal tube stop—steel teeth on the collet grip tubing for leakproof connections. To disconnect, press collet against fitting body to disengage steel teeth—tubing slides right out.<br /><br />Fittings work with semi-rigid plastic (polypropylene, polyethylene, nylon) and soft metal (brass, copper, mild steel) tubing.<br />Adapters (sold separately) let you permanently modify basic fittings for customized installations.