GF Signet P526-2007 316 SS Socket-Weld Mini-Tap Fittings; 3/4" Pipe ID
供应商: MASTERFLEX PREMIUM
Installation fittings for your GF® Signet Paddle Wheel Flowmeters.
- 316 SS Socket-Weld Mini-Tap Fittings for Metalex™ sensors only
- Includes cap kit
Fittings are precision crafted to ensure proper sensor insertion depth and accurate flow measurement. Pipe fitting sizes range from 1/2" to 1" in diameter.<br />Required system components are the flow sensor, installation fitting, and monitor or controller, all sold separately.