Masterflex® Transfer Tubing, PVC, 1/4" ID x 1/2" OD; 50 Ft
供应商: MASTERFLEX PREMIUM
VWR Item#MFLB96605-05
Your Organization's Price:
正在获取
Stop oxidation and bacteria problems.
  • Ideal for general-purpose, single-use, and extended fluid path applications
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimise downtime
  • Regulatory approvals to match your unique application
  • Tubing size: 1/4" ID x 1/2" OD
Cole-Parmer PVC 泵管挠性很强,管身透明,抗氧化,细菌和多种化学品。该泵管管符合FDA (21 CFR 175.300) 规范,并达到 USP VI 级和 USDA 要求,以及 3A 无菌标准,非常适合食品和饮料加工,以及制药/生物技术应用。这种泵管的工作温度范围为 -41 至 165 F (-41 至 74 C)。 可以使用环氧乙烷或伽马射线消毒。注意:不建议使用高压灭菌器消毒。