Masterflex® L/S® Easy-Load® II Pump Head for Precision Tubing, PPS Housing, SS Rotor
供应商: MASTERFLEX PROPRIETARY
自动固位装置可将泵管牢固固定到位,无需人工调节,—只需装载泵管,然后启动泵即可。
- 四辊轴马达提高了泵头的压力性能和机械稳定性,同时降低了流动脉动。
- 改良的咬合床符合几何学原理,延长了泵管使用寿命。
- 聚苯硫醚(PPS)外壳,不锈钢(SS)和冷轧钢(CRS)两种转子型号可选—CRS转子的泵头价格实惠,而SS抗化学性更佳且更耐用。
- 77201-60型泵头咬合可调节,您可放松咬合以延长泵管使用寿命,或调紧咬合以获得更高的压力。
- 每一种兼容精密泵管的泵头都可兼容L/S® 13, L/S® 14, L/S® 16, L/S® 25, L/S® 17 和L/S® 18泵管。
- 流速范围可达0.06~2300 mL/min (1 ~ 600 rpm)。
- Rotor is constructed of stainless steel (SS) for enhanced durability
如需叠连多个泵头(最多可达驱动器的规定极限数额),请在“附件”部分单独订购 2、3 及 4 通道配件。当叠连 2 个 Easy-Load II 泵头时,辊子会偏移 45°— 以合并 2 个叠连泵头的出口,从而降低 95% 的流动脉动。
This pump head requires an L/S drive equipped with open-head sensor functionality or the use of the 77097-00 open-head sensor retrofit controller.
<b>Delivery information: </b>Includes single-channel mounting hardware. For stacking multiple heads (up to the specified limits of your drive) order two- three- or four-channel hardware separately.
Includes
<b>Delivery information: </b>Includes single-channel mounting hardware. For stacking multiple heads (up to the specified limits of your drive) order two- three- or four-channel hardware separately.
Key Features
- 四辊轴马达提高了泵头的压力性能和机械稳定性,同时降低了流动脉动。
- 改良的咬合床符合几何学原理,延长了泵管使用寿命。
- 聚苯硫醚(PPS)外壳,不锈钢(SS)和冷轧钢(CRS)两种转子型号可选—CRS转子的泵头价格实惠,而SS抗化学性更佳且更耐用。
- 77201-60型泵头咬合可调节,您可放松咬合以延长泵管使用寿命,或调紧咬合以获得更高的压力。
- 每一种兼容精密泵管的泵头都可兼容L/S® 13, L/S® 14, L/S® 16, L/S® 25, L/S® 17 和L/S® 18泵管。
- 流速范围可达0.06~2300 mL/min (1 ~ 600 rpm)。
- Rotor is constructed of stainless steel (SS) for enhanced durability
More About This Item
<br>自动固位装置可将泵管牢固固定到位,无需人工调节,—只需装载泵管,然后启动泵即可。<br><br>如需叠连多个泵头(最多可达驱动器的规定极限数额),请在“附件”部分单独订购 2、3 及 4 通道配件。当叠连 2 个 Easy-Load II 泵头时,辊子会偏移 45°— 以合并 2 个叠连泵头的出口,从而降低 95% 的流动脉动。<br><br>This pump head requires an L/S drive equipped with open-head sensor functionality or the use of the 77097-00 open-head sensor retrofit controller.<br>