Masterflex® Syringe Pump, Infusion and Withdrawal, Programmable Touchscreen Control; 100/240 VAC
供应商: MASTERFLEX PROPRIETARY
VWR Item#MFLA74905-54
Your Organization's Price:
正在获取
通过界面友好的触摸屏显示器控制流速,范围为 5 pL/min 到 200 mL/min。
  • 全触摸屏,方便您快速创建配置以备随后调用
  • 可侧面放置泵体以节省空间,触摸屏显示器将自动旋转
  • 精度为 ±0.355%,再现性为 ±0.05%
  • 可通过内置的 RS-485 连接菊链串接多达 99 台泵
  • 前面板的 LED 指示灯可直观提示泵正在运行中
  • 兼容所有类型的注射器 — 玻璃、塑料或不锈钢,容量 0.5 µL 至 140 mL
  • 全金属机箱具备优异的噪声隔离和防震功能,提高可靠性
These Masterflex® touch-screen syringe pumps offer user-friendly programming and control. All pumps feature an accuracy of ±0.355% and a reproducibility of ±0.05% along with output options RS-232, RS-485, USB, and 15 pin D-sub connector.
<B>Model 74905-54</B> is a programmable version of the 74905-52 pump. It is programmed to accept up to 800 user-defined steps—up to 40 programs with 20 steps each can be stored in memory.
<b>Delivery information: </b>6 ft (1.8 m) 美国标准电源线。