Masterflex® Analog Gear Pump System, 0.017 mL/rev, PTFE Seal; 115 VAC
供应商: MASTERFLEX PROPRIETARY
VWR Item#MFLA74013-72
Your Organization's Price:
正在获取
Simple variable speed control handles many fluid handling applications.
  • 100:1 speed-control range controls flow over a wide range
  • Simple operation with single-turn speed control
  • Separate power switch enables you to maintain speed setting when turning drive on/off; front-panel LED indicates power on
  • Remote control of start/stop via DB9 connector on back of drive
  • Flow rate of 0.017 mL/rev
Compact, cost-effective drive offers simple variable speed control for many fluid handling applications from general transfer to filtration to chemical injection. Conveniently coupled with an A-mount gear pump head for pulseless fluid flow.