Masterflex® Fitting, Polypropylene, Straight, Female Luer to Bottom Sealing Thread, 1/4-28 UNF; 25/PK
供应商: MASTERFLEX PROPRIETARY
VWR Item#MFLA45508-76
Your Organization's Price:
正在获取
Easily transition between luer fittings and UNF or NPT threads.
  • Repackaged in an ISO 9001 facility in an ISO Class 7 Cleanroom
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimise downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
  • Connection Type: Female Luer to Bottom Sealing Threaded, 1/4-28 UNF
These adapters transition between luer fittings and UNF or NPT threads. Both male and female luer connections are available. All threaded connections are female thread.