Masterflex® USB Cable; Type A Male to Type B Male; Beige, 10 ft L
供应商: MASTERFLEX PROPRIETARY
VWR Item#MFLA22050-60
Your Organization's Price:
正在获取
Optimize performance of your Masterflex® pump.
  • Suitable for use with Masterflex® L/S® 07551-series computer-compatible drives
使用这一线缆将Masterflex® L/S® 07551系列兼容计算机的驱动器连接到您的电脑。