Masterflex® L/S® Analog Fixed-Speed Compact Drive, 100 rpm; 12 VDC
供应商: MASTERFLEX PROPRIETARY
直流驱动器体积小,适合现场使用和OEM应用。
- 钢制马达外壳,涂有环氧树脂涂层,耐化学腐蚀,防护等级为IP52—防尘、防轻微水喷。
- 通过改变输入电压调节驱动器转速;将电源正负极对调则可使马达反向运行。完全通过供电电源来控制驱动器工作状态。
- 坚固可靠的马达能够全天候连续稳定运行。
- 电源线为三芯16号导线,长12英寸(30厘米),裸线头。使用12V或24V直流电源供电。
- Accepts up to 2 pump heads
这款驱动器非常适合用于OEM应用。驱动器拥有带环氧树脂涂层的马达,裸线电源线,安装架,能够轻松安装到您的设备上。
†Drives that accept only one pump head cannot be used with the High-Performance or PTFE-Tubing pump heads due to insufficient torque.