Masterflex® Ismatec® Pump Tubing, 2-Stop, Santoprene®, 0.89 mm ID; 12/PK
供应商: MASTERFLEX PROPRIETARY
Choose the size and formulation to suit your flow rate and application needs.
- Genuine Ismatec® tubing undergoes robust testing to provide optimal performance / pump life hours with peristaltic pump equipment
- Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
- Global market leader in range of formulations for most extensive chemical compatibility
- Single-use custom lengths and configurations available to minimise downtime
- Regulatory approvals to match your unique application
- Tubing Inner Diameter: 0.89 mm
Compatible with Masterflex® L/S® Multichannel pump head, small cartridges