Masterflex® Variable-Area Flowmeter with Valve, Correlated Reading, PTFE Housing and Fittings, 65-mm Scale; 159 mL/min Air
供应商: MASTERFLEX PROPRIETARY
VWR Item#MFLA03216-60
Your Organization's Price:
正在获取