Masterflex® L/S® High-Performance Precision Pump Tubing, C-Flex®, L/S 36; 25 ft
供应商: MASTERFLEX PROPRIETARY
既有硅胶管的生物相容性,又有类似Tygon®的化学相容性。
- 极低的蛋白结合度
- 可热封、可焊接
- 经济实惠
- Tubing Size: L/S 36
<B>用途</B>:要求高性能和长寿命的通用实验室用途。<br /><B>特点</B>:卓越的抗拉和撕裂强度;良好的生物兼容性;低透气性;表面平滑。白色不透明。
See 96123-15 for the irradiated version of this tubing.