Masterflex® I/P® High-Performance Precision Pump Tubing, Versilon™ A-60-N, I/P 89; 25 ft
供应商: MASTERFLEX PROPRIETARY
VWR Item#MFLA06404-89
Your Organization's Price:
正在获取
Outlast and outperform other tubing with 10,000 hours of tubing life.
  • Genuine Masterflex® tubing undergoes robust testing to provide optimal performance/pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy, and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimise downtime
  • Regulatory Approvals to match your unique application
  • Opaque beige color protects light and UV sensitive samples
  • I/P® 89 tubing has a 5/8” (15.88 mm) inner diameter
  • Accepts 5/8” hose barb for tubing connection
<B>应用</B>: 应用要求耐化学,耐热,耐臭氧环境,耐紫外线.<br /><B>特点</B>: 不传导,热密封,抗老化,抗氧化,良好的耐酸/碱性. 不透明,黑色。