Masterflex® L/S® 2-Stop High-Performance Precision Pump Tubing, Tygon® S3™ E-LFL, L/S 24; 4/Pk
供应商: MASTERFLEX PROPRIETARY
VWR Item#MFLA06447-24
Your Organization's Price:
正在获取
和Masterflex® L/S®多通道泵头搭配使用。
  • Genuine Masterflex® tubing undergoes robust testing to provide optimal performance / pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Regulatory approvals to match your unique application
  • Tubing Size: L/S 24
For use with Masterflex® L/S® multichannel pump heads.