WHEATON® Bottles, Wide Neck, with Screw Cap
供应商: DWK LIFE SCIENCES
W216947 W216946
WHEAW216947EA 0 CNY
WHEAW216947 WHEAW216946
WHEATON® Bottles, Wide Neck, with Screw Cap
玻璃瓶
Amber, USP Type III soda-lime glass.
  • Easy removal of contents
  • White polypropylene screw cap
  • Hazardous communication label included with each case of bottles allows for content ID and safety notification
  • Tamper-evident seal included with each case of bottles keeps bottle sealed which protecting contents from accidental misuse
Ideal for storage of light-sensitive samples or dry powders.