Syringeless filters, Mini-UniPrep™, Whatman™
供应商: CYTIVA WHATMAN
UN203NPUORG UN203NPEORG UN503NPUORG UN203NPUGMF UN203NPEAQU UN203NPENYL UN503NPEAQU UN503NPENYL UN503NPEORG UN203APEAQU UN203APUORG UN203NPUAQU UN203NPUDPP UN203NPUNYL UN503NPUAQU UN503NPUDPP UN503NPUNYL US203NPEAQU US203NPEORG US203NPUAQU US203NPUGMF US203NPUNYL US203NPUORG US203NPUPES US503NPEAQU US503NPENYL US503NPEORG US503NPUAQU US503NPUORG US503NPUGMF US503NPUDPP UN203NPERC UN203NPURC UN503NPERC US203APUNYL
WHATUN203NPUORGEA 0 CNY
WHATUN203NPUORG WHATUN203NPEORG WHATUN503NPUORG WHATUN203NPUGMF WHATUN203NPEAQU WHATUN203NPENYL WHATUN503NPEAQU WHATUN503NPENYL WHATUN503NPEORG WHATUN203APEAQU WHATUN203APUORG WHATUN203NPUAQU WHATUN203NPUDPP WHATUN203NPUNYL WHATUN503NPUAQU WHATUN503NPUDPP WHATUN503NPUNYL WHATUS203NPEAQU WHATUS203NPEORG WHATUS203NPUAQU WHATUS203NPUGMF WHATUS203NPUNYL WHATUS203NPUORG WHATUS203NPUPES WHATUS503NPEAQU WHATUS503NPENYL WHATUS503NPEORG WHATUS503NPUAQU WHATUS503NPUORG WHATUS503NPUGMF WHATUS503NPUDPP WHATUN203NPERC WHATUN203NPURC WHATUN503NPERC WHATUS203APUNYL
Syringeless filters, Mini-UniPrep™, Whatman™
过滤器 非针式滤器
The Whatman Mini-UniPrep Syringeless Filters combine four products into one syringeless filter solution for efficient chromatography sample preparation for a broad range of applications. They are built for fast and easy high performance liquid chromatography/ultra high performance liquid chromatography sample preparation and analysis.
  • All-in-one filtration device for quick and cost-effective chromatography sample processing
  • Eliminates the need for separate syringes, syringe membrane filters, vials, and septa
  • Designed for use with an UHPLC or HPLC
More Product Information