Syringe filters, nylon, Puradisc™, Whatman™
供应商: CYTIVA WHATMAN
6750-2502 6750-2504 6751-2502 6751-2504 6789-1301 6789-1302 6789-1304 6790-1302 6790-1304 6750-2510 6752-2504 6753-2502 6753-2504 6753-2510 6768-1302 6768-1304
WHAT6750-2502EA 0 CNY
WHAT6750-2502 WHAT6750-2504 WHAT6751-2502 WHAT6751-2504 WHAT6789-1301 WHAT6789-1302 WHAT6789-1304 WHAT6790-1302 WHAT6790-1304 WHAT6750-2510 WHAT6752-2504 WHAT6753-2502 WHAT6753-2504 WHAT6753-2510 WHAT6768-1302 WHAT6768-1304
Syringe filters, nylon, Puradisc™, Whatman™
过滤器 注射器过滤器
Whatman Puradisc syringe filters from Cytiva's business combine premium quality with economic efficiency. They are well suited for rapid, routine syringe filtration of samples up to 100 ml.
  • Eight different material options
  • Choice of 4 mm, 13 mm, and 30 mm syringe filter diameter to suit sample volume
  • Range of pore sizes for retaining a variety of particles
  • Optional sterile, medical-grade blister pack
  • Optional tube-tip format for accurate dispensing into a micro-vial
  • Recyclable after use
Puradisc syringe filters cater to a wide variety of sample typesMore Product Information