Cellulose chromatography papers, Whatman™
供应商: CYTIVA WHATMAN
3001-604 3001-614 3001-640 3001-652 3001-653 3001-672 3001-681 3001-845 3001-861 3001-878 3001-917 3001-931 3001-964 3002-917 3003-917 3004-917 3017-915 3030-335 3030-614 3030-662 3030-672 3030-675 3030-681 3030-690 3030-700 3030-704 3030-861 3030-917 3030-931 3031-915 10382461 10382514 10382562 10382581 1030-024 1030-025 3030-392 3017-8793 3030-6188 3454-651 3020-917 3030-6461 3030-221 3030-6185 3030-6132 3030-347 3017-621 3030-153 3001-651 3002-911 3004-614 3017-917 3030-6189 3030-866 3031-681 3017-820 3017-8823 3031-901
WHAT3001-604EA 0 CNY
WHAT3001-604 WHAT3001-614 WHAT3001-640 WHAT3001-652 WHAT3001-653 WHAT3001-672 WHAT3001-681 WHAT3001-845 WHAT3001-861 WHAT3001-878 WHAT3001-917 WHAT3001-931 WHAT3001-964 WHAT3002-917 WHAT3003-917 WHAT3004-917 WHAT3017-915 WHAT3030-335 WHAT3030-614 WHAT3030-662 WHAT3030-672 WHAT3030-675 WHAT3030-681 WHAT3030-690 WHAT3030-700 WHAT3030-704 WHAT3030-861 WHAT3030-917 WHAT3030-931 WHAT3031-915 WHAT10382461 WHAT10382514 WHAT10382562 WHAT10382581 WHAT1030-024 WHAT1030-025 WHAT3030-392 WHAT3017-8793 WHAT3030-6188 WHAT3454-651 WHAT3020-917 WHAT3030-6461 WHAT3030-221 WHAT3030-6185 WHAT3030-6132 WHAT3030-347 WHAT3017-621 WHAT3030-153 WHAT3001-651 WHAT3002-911 WHAT3004-614 WHAT3017-917 WHAT3030-6189 WHAT3030-866 WHAT3031-681 WHAT3017-820 WHAT3017-8823 WHAT3031-901
Cellulose chromatography papers, Whatman™
过滤器 滤纸 Chromatography Paper
Whatman chromatography papers are widely used across the world, reflecting their purity, high quality and consistency. They are made from specially selected cotton cellulose and rigorously quality controlled to ensure uniformity within the grade.
  • Pure cellulose produced from the highest quality cotton linters, with no additives, ensures that no contamination will occur during the transfer steps
  • Manufactured and tested specifically for chromatographic techniques to ensure the wicking capability and uniformity of capillary action that is important in
More Product Information