Weighing papers, grade 2122, Whatman™
供应商: CYTIVA WHATMAN
10347890 10347893
WHAT10347890EA 0 CNY
WHAT10347890 WHAT10347893
Weighing papers, grade 2122, Whatman™
称量纸
These Grade 2122 weigh paper sheets reliably allow samples to be transferred to scales without adding unwanted substances that could impact analytical results.
  • Smooth surface allows samples to slide off with ease and not be retained on paper
  • Shiny pergamyne paper allows visual determination that all sample has been removed
  • Extremely thin (0,03 mm) and lightweight (40 g/m²) for safe and secure sample transport
  • No glue or additives (residue-free)
This paper is not low in nitrogen. Therefore, it should not be used for Kjeldahl analysis.