Qualitative filter papers, standard grades, grade 593 ¹/₂, Whatman™
供应商: CYTIVA WHATMAN
10311447 10311451
WHAT10311447EA 0 CNY
WHAT10311447 WHAT10311451
Qualitative filter papers, standard grades, grade 593 ¹/₂, Whatman™
过滤器 滤纸 Qualitative Filter Paper
These cellulose filters are used in qualitative analytical techniques to determine and identify materials. A standard grade filter paper for fine precipitates.
  • Increased filter surface area increases total loading capacity and decreases overall filtration time
  • Reduced contact between filter and funnel maintains flow rate
  • Eliminated need to quadrant-fold filter circles to fit conical funnels saves time in repetitive or multiple analyses
  • Prepleated filters have the same technical properties as the respective filter circles
Prepleated qualitative filtersMore Product Information