Grade 3 qualitative filter papers, Whatman™
供应商: CYTIVA WHATMAN
1003-055 1003-070 1003-090 1003-110 1003-125 1003-150 1003-185 1003-240 1003-320 1003-917 1003-323
WHAT1003-055EA 0 CNY
WHAT1003-055 WHAT1003-070 WHAT1003-090 WHAT1003-110 WHAT1003-125 WHAT1003-150 WHAT1003-185 WHAT1003-240 WHAT1003-320 WHAT1003-917 WHAT1003-323
Grade 3 qualitative filter papers, Whatman™
过滤器 滤纸 Qualitative Filter Paper
A standard grade filter used in qualitative analytical techniques to determine and identify materials. For use in büchner funnels.
  • Thickness of 6 µm
Prepleated qualitative filters are also available, which give improved flow rate and increased loading capacity compared to equivalent flat filters.
Double the thickness of Grade 1 with still finer particle retention and excellent loading capacity; more precipitate can be held without clogging. The extra thickness gives increased wet strength and makes this grade highly suitable for use in BüchnerMore Product Information