VWR® Talon®, Support stands, heavy duty
供应商: VWR BRAND PRIVATE LABEL USA
97035-510 97035-516 97035-514
VWRU97035-510EA 5731 CNY
VWRU97035-510 VWRU97035-516 VWRU97035-514
VWR® Talon®, Support stands, heavy duty
支撑架
Heavy-duty support stands permit cylindrical vessels up to 30,5 cm (12") in diameter to sit close to the support rod. Ideal for mounting stirring motors for heavy equipment.
  • Base is constructed of cast iron with a black enamel finish
  • Each of the four corners rests on a rubber cushion
  • Includes a 16 mm (⁵/₈") diameter support rod
  • Extra stainless steel support rods are available separately