VWR®, Filter forceps
供应商: VWR BRAND PRIVATE LABEL USA
89259-954
VWRU89259-954EA 103 CNY
VWRU89259-954
VWR®, Filter forceps
镊子
These stainless steel forceps with black PP non-slip finger grips are ideal for handling membranes.