VWR®, Refrigerated circulating baths, 240 V
供应商: VWR BRAND PRIVATE LABEL USA
89202-996 89203-024
VWRU89202-996EA 165772 CNY
VWRU89202-996 VWRU89203-024
VWR®, Refrigerated circulating baths, 240 V
循环水浴 闭式循环系统 Cooling Circulators
VWR® Refrigerated Circulating Baths offer precise temperature control, at temperature ranges from -40 to 200°C (depending on the controller).
  • Easy-to-view LCD display
  • Precise temperature control
Available in a wide range of sizes, with three controller options, these refrigerated circulating baths are ideal for many applications.
The large LCD display on the MX temperature controller is easily viewed across the lab. The Advanced Digital temperature controller offers a touch-pad, while the Advanced Programmable Temperature Controller includes a fullMore Product Information